1,4'-Bipiperidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Piperidinopiperidine

AS20665G25 AS20665G5
ACTIAS20665G25EA 73 CHF
ACTIAS20665G25 ACTIAS20665G5
1,4'-Bipiperidin
1,4'-Bipiperidin
Formel: C₁₀H₂₀N₂
MW: 168,28 g/mol
MDL Number: MFCD00006475
CAS-Nummer: 4897-50-1
EINECS: 225-522-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR