N(α)-Fmoc-L-tryptophan

Lieferant: Molekula

MOLE29625065-25GEA 0 CHF
MOLE29625065-25G
N(α)-Fmoc-L-tryptophan
N(α)-Fmoc-L-tryptophan
Formel: C₂₆H₂₂N₂O₄
MW: 426,5 g/mol
MDL Number: MFCD00037126
CAS-Nummer: 35737-15-6
EINECS: 252-706-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR