N(α)-Fmoc-D-tryptophan

Lieferant: BACHEM BIOCHEMICA

BACE4003200.0025EA 490 CHF
BACE4003200.0025 BACE4003200.0005
N(α)-Fmoc-D-tryptophan
N(α)-Fmoc-D-tryptophan
Formel: C₂₆H₂₂N₂O₄
MW: 426,46 g/mol
MDL Number: MFCD00062954
CAS-Nummer: 86123-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR