6-Piperidino-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS13026G1 AS13026G5
ACTIAS13026G1EA 0 CHF
ACTIAS13026G1 ACTIAS13026G5
6-Piperidino-3-pyridinboronsäure
6-Piperidino-3-pyridinboronsäure
Formel: C₁₀H₁₅BN₂O₂
MDL Number: MFCD06801705
CAS-Nummer: 1002129-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR