2,4-Dichlor-7-methylchinazolin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS2590G1EA 639 CHF
ACTIAS2590G1 ACTIAS2590G5
2,4-Dichlor-7-methylchinazolin
2,4-Dichloro-7-methylquinazoline
Formel: C₉H₆Cl₂N₂
MW: 213,0633 g/mol
MDL Number: MFCD11858254
CAS-Nummer: 25171-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR