6-Brom-4-chlor-2-methylchinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS30759G1 AS30759G5
ACTIAS30759G1EA 0 CHF
ACTIAS30759G1 ACTIAS30759G5
6-Brom-4-chlor-2-methylchinazolin
6-Bromo-4-chloro-2-methylquinazoline
Formel: C₉H₆BrClN₂
MW: 257,5143 g/mol
MDL Number: MFCD06657210
CAS-Nummer: 351426-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR