1,2-Dimethyl-5-(tributylstannyl)imidazol

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS50710G1 AS50710G5
ACTIAS50710G1EA 0 CHF
ACTIAS50710G1 ACTIAS50710G5
1,2-Dimethyl-5-(tributylstannyl)imidazol
1,2-Dimethyl-5-(tributylstannyl)imidazol
Formel: C₁₇H₃₄N₂Sn
MDL Number: MFCD11109395
CAS-Nummer: 86051-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR