5-Nitro-2-pyridincarbonitril

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS31654G5EA 0 CHF
ACTIAS31654G5 ACTIAS31654G1
5-Nitro-2-pyridincarbonitril
5-Nitro-2-pyridincarbonitril
Formel: C₆H₃N₃O₂
MW: 149,1069 g/mol
MDL Number: MFCD07368184
CAS-Nummer: 100367-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR