4-Chlor-N-methylpicolinamid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloro-N-methylpyridine-2-carboxamide

AS31795G1 AS31795G5
ACTIAS31795G1EA 0 CHF
ACTIAS31795G1 ACTIAS31795G5
4-Chlor-N-methylpicolinamid
4-Chlor-N-methylpicolinamid
Formel: C₇H₇ClN₂O
MW: 170,60 g/mol
MDL Number: MFCD02185921
CAS-Nummer: 220000-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR