4-Benzyloxy-3-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51249G1 AS51249G5
ACTIAS51249G1EA 0 CHF
ACTIAS51249G1 ACTIAS51249G5
4-Benzyloxy-3-methoxyphenylboronsäure
4-Benzyloxy-3-methoxyphenylboronsäure
Formel: C₁₄H₁₅BO₄
MDL Number: MFCD09027252
CAS-Nummer: 243990-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR