Thiophenol auf Polystyrol

Lieferant: BIOTAGE

Synonyms: Mercaptobenzene, Phenyl mercaptan, Benzenethiol

800500 800273 800274 800275 800310
BITA800500EA 0 CHF
BITA800500 BITA800273 BITA800274 BITA800275 BITA800310
Thiophenol auf Polystyrol
Thiophenol
Formel: C₆H₆S
MDL Number: MFCD00004826
CAS-Nummer: 108-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR