1-Methylimidazol-5-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS1414G1 AS1414G5
ACTIAS1414G1EA 0 CHF
ACTIAS1414G1 ACTIAS1414G5
1-Methylimidazol-5-carbaldehyd
1-Methylimidazol-5-carbaldehyd
Formel: C₅H₆N₂O
MDL Number: MFCD00030439
CAS-Nummer: 39021-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR