5-Brompicolinamid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromopyridine-2-carboxamide
AS35418G1
AS35418G5
ACTIAS35418G1EA
0
CHF
ACTIAS35418G1
ACTIAS35418G5
5-Brompicolinamid
5-Brompicolinamid
Formel:
C₆H₅BrN₂O MW: 201.0200 g/mol |
MDL Number:
MFCD04066715 CAS-Nummer: 90145-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...