3-Hydroxytetrahydropyran

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Hydroxy-2H-pyran, Tetrahydro-2H-pyran-3-ol

AS32134G1 AS32134G5
ACTIAS32134G1EA 0 CHF
ACTIAS32134G1 ACTIAS32134G5
3-Hydroxytetrahydropyran
3-Hydroxytetrahydropyran
Formel: C₅H₁₀O₂
MW: 102,1300 g/mol
MDL Number: MFCD00219387
CAS-Nummer: 19752-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR