4'-Ethylbiphenyl-4-ylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4'-Ethyl-[1,1'-biphenyl]-4-yl)boronic acid

AS24240G1 AS24240G25 AS24240G5
ACTIAS24240G1EA 59 CHF
ACTIAS24240G1 ACTIAS24240G25 ACTIAS24240G5
4'-Ethylbiphenyl-4-ylboronsäure
4'-Ethylbiphenyl-4-ylboronsäure
Formel: C₁₄H₁₅BO₂
MW: 226,0800 g/mol
MDL Number: MFCD04038068
CAS-Nummer: 153035-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR