3-(tert-Butoxycarbonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-(tert-Butoxycarbonyl)benzeneboronic acid, 3-tert-Butoxycarbonylphenylboronic acid

AS50280G1 AS50280G5
ACTIAS50280G1EA 0 CHF
ACTIAS50280G1 ACTIAS50280G5
3-(tert-Butoxycarbonyl)phenylboronsäure
3-(tert-Butoxycarbonyl)phenylboronsäure
Formel: C₁₁H₁₅BO₄
MW: 222,05 g/mol
MDL Number: MFCD01630855
CAS-Nummer: 220210-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR