4-(2-Fluor-4-nitrophenyl)morpholin

Lieferant: ACTIVATE SCIENTIFIC

AS21822G25 AS21822G5
ACTIAS21822G25EA 0 CHF
ACTIAS21822G25 ACTIAS21822G5
4-(2-Fluor-4-nitrophenyl)morpholin
4-(2-Fluor-4-nitrophenyl)morpholin
Formel: C₁₀H₁₁FN₂O₃
MDL Number: MFCD03138381
CAS-Nummer: 2689-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR