2,4-Dichlor-8-methoxychinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS2577G1 AS2577G5
ACTIAS2577G1EA 0 CHF
ACTIAS2577G1 ACTIAS2577G5
2,4-Dichlor-8-methoxychinazolin
2,4-Dichloro-8-methoxyquinazoline
Formel: C₉H₆Cl₂N₂O
MDL Number: MFCD09954882
CAS-Nummer: 61948-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR