2,4-Dichlor-7-methoxychinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS2594 AS2594G1 AS2594G5
ACTIAS2594EA 0 CHF
ACTIAS2594 ACTIAS2594G1 ACTIAS2594G5
2,4-Dichlor-7-methoxychinazolin
2,4-Dichloro-7-methoxyquinazoline
Formel: C₉H₆Cl₂N₂O
MDL Number: MFCD09954881
CAS-Nummer: 62484-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR