2,4-Dichlor-7-fluorchinazolin
Lieferant: ACTIVATE SCIENTIFIC
AS2588G1
AS2588G5
ACTIAS2588G1EA
0
CHF
ACTIAS2588G1
ACTIAS2588G5
2,4-Dichlor-7-fluorchinazolin
2,4-Dichloro-7-fluoroquinazoline
Formel:
C₈H₃Cl₂FN₂ MW: 217,0272 g/mol |
MDL Number:
MFCD09954885 CAS-Nummer: 174566-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...