4-Chlorchinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS20236G1 AS20236G25 AS20236G5
ACTIAS20236G1EA 0 CHF
ACTIAS20236G1 ACTIAS20236G25 ACTIAS20236G5
4-Chlorchinazolin
4-Chloroquinazoline
Formel: C₈H₅ClN₂
MW: 164,5900 g/mol
MDL Number: MFCD00228682
CAS-Nummer: 5190-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR