4,5-Dichlorchinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS20237G1 AS20237G5
ACTIAS20237G1EA 154 CHF
ACTIAS20237G1 ACTIAS20237G5
4,5-Dichlorchinazolin
4,5-Dichloroquinazoline
Formel: C₈H₄Cl₂N₂
MW: 199,0400 g/mol
MDL Number: MFCD09909637
CAS-Nummer: 2148-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR