3-Chlor-5-(trifluormethyl)picolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Chloro-5-(trifluoromethyl)-2-pyridinecarboxylic acid, 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 3-Chloro-5-(trifluoromethyl)picolinic acid

AS30883G1 AS30883G5
ACTIAS30883G1EA 0 CHF
ACTIAS30883G1 ACTIAS30883G5
3-Chlor-5-(trifluormethyl)picolinsäure
3-Chlor-5-(trifluormethyl)picolinsäure
Formel: C₇H₃ClF₃NO₂
MW: 225,60 g/mol
MDL Number: MFCD00277482
CAS-Nummer: 80194-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR