2-Brom-6-iodbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS36463G1 AS36463G5
ACTIAS36463G1EA 0 CHF
ACTIAS36463G1 ACTIAS36463G5
2-Brom-6-iodbenzonitril
2-Brom-6-iodbenzonitril
Formel: C₇H₃BrIN
MW: 307,9139 g/mol
MDL Number: MFCD16619725
CAS-Nummer: 1245648-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR