2-Brom-4-(trifluormethoxy)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32632G1 AS32632G5
ACTIAS32632G1EA 0 CHF
ACTIAS32632G1 ACTIAS32632G5
2-Brom-4-(trifluormethoxy)benzonitril
2-Brom-4-(trifluormethoxy)benzonitril
Formel: C₈H₃BrF₃NO
MW: 266,0147 g/mol
MDL Number: MFCD13185874
CAS-Nummer: 1214334-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR