(2-Chlorphenyl)acetamid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(2-Chlorophenyl)acetamide

AS24769G1 AS24769G25 AS24769G5
ACTIAS24769G1EA 44 CHF
ACTIAS24769G1 ACTIAS24769G25 ACTIAS24769G5
(2-Chlorphenyl)acetamid
(2-Chlorphenyl)acetamid
Formel: C₈H₈ClNO
MW: 169.6100 g/mol
MDL Number: MFCD00176724
CAS-Nummer: 10268-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR