Methyl-6-brompicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 6-bromopicolinate, Methyl 6-bromopyridine-2-carboxylate, 6-Bromopyridine-2-carboxylic acid methyl ester, 6-Bromopicolinic acid methyl ester

AS31614G1 AS31614G5
ACTIAS31614G1EA 0 CHF
ACTIAS31614G1 ACTIAS31614G5
Methyl-6-brompicolinat
Methyl-6-brompicolinat
Formel: C₇H₆BrNO₂
MW: 216,03 g/mol
MDL Number: MFCD06203934
CAS-Nummer: 26218-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR