2-Amino-4-chlor-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS1878G1 AS1878G5
ACTIAS1878G1EA 0 CHF
ACTIAS1878G1 ACTIAS1878G5
2-Amino-4-chlor-3-nitropyridin
2-Amino-4-chloro-3-nitropyridine
Formel: C₅H₄ClN₃O₂
MDL Number: MFCD04116097
CAS-Nummer: 6980-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR