(5-Fluor-2-nitrophenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Fluoro-2-nitrophenylacetic acid

AS33380G1 AS33380G10 AS33380G5
ACTIAS33380G1EA 0 CHF
ACTIAS33380G1 ACTIAS33380G10 ACTIAS33380G5
(5-Fluor-2-nitrophenyl)essigsäure
(5-Fluor-2-nitrophenyl)essigsäure
Formel: C₈H₆FNO₄
MDL Number: MFCD07787601
CAS-Nummer: 29640-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR