5,6-Dichlor-2-methylbenzimidazol

Lieferant: ACTIVATE SCIENTIFIC

AS35228G1 AS35228G5
ACTIAS35228G1EA 0 CHF
ACTIAS35228G1 ACTIAS35228G5
5,6-Dichlor-2-methylbenzimidazol
5,6-Dichloro-2-methylbenzimidazole
Formel: C₈H₆Cl₂N₂
MW: 201,0500 g/mol
MDL Number: MFCD00276973
CAS-Nummer: 6478-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR