2-Methylisonicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methylpyridine-4-carboxylic acid

AS1096G1 AS1096G5
ACTIAS1096G1EA 0 CHF
ACTIAS1096G1 ACTIAS1096G5
2-Methylisonicotinsäure
2-Methylisonicotinsäure
Formel: C₇H₇NO₂
MW: 137,14 g/mol
MDL Number: MFCD00513435
CAS-Nummer: 4021-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR