2-Amino-3,5-dibrom-4-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,5-Dibromo-4-methylpyridin-2-amine

AS1565G1 AS1565G5
ACTIAS1565G1EA 0 CHF
ACTIAS1565G1 ACTIAS1565G5
2-Amino-3,5-dibrom-4-methylpyridin
2-Amino-3,5-dibromo-4-methylpyridine
Formel: C₅H₄Br₂N₂
MW: 265.9300 g/mol
MDL Number: MFCD01646065
CAS-Nummer: 3430-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR