4-(Methylcarbamoyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(Methylcarbamoyl)benzeneboronic acid, 4-(N-Methylaminocarbonyl)phenylboronic acid

AS32713G1 AS32713G5
ACTIAS32713G1EA 0 CHF
ACTIAS32713G1 ACTIAS32713G5
4-(Methylcarbamoyl)phenylboronsäure
4-(Methylcarbamoyl)phenylboronsäure
Formel: C₈H₁₀BNO₃
MW: 178.98 g/mol
MDL Number: MFCD03788427
CAS-Nummer: 121177-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR