4-(Ethylcarbamoyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(N-Ethylaminocarbonyl)phenylboronic acid

AS51345G1 AS51345G5
ACTIAS51345G1EA 0 CHF
ACTIAS51345G1 ACTIAS51345G5
4-(Ethylcarbamoyl)phenylboronsäure
4-(Ethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
MW: 193,0100 g/mol
MDL Number: MFCD03411954
CAS-Nummer: 850568-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR