4-(Dimethylcarbamoyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-(Dimethylcarbamoyl)phenyl)boronic acid, 4-(Dimethylcarbamoyl)benzeneboronic acid, 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid

AS50328G1 AS50328G5
ACTIAS50328G1EA 0 CHF
ACTIAS50328G1 ACTIAS50328G5
4-(Dimethylcarbamoyl)phenylboronsäure
4-(Dimethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
MW: 193,01 g/mol
MDL Number: MFCD02258943
CAS-Nummer: 405520-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR