4-(Diethylcarbamoyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(Diethylcarbamoyl)benzeneboronic acid, 4-(N,N-Diethylaminocarbonyl)phenylboronic acid

AS51239G1 AS51239G5
ACTIAS51239G1EA 0 CHF
ACTIAS51239G1 ACTIAS51239G5
4-(Diethylcarbamoyl)phenylboronsäure
4-(Diethylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
MW: 221,06 g/mol
MDL Number: MFCD03411949
CAS-Nummer: 389621-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR