3-(3-Fluorphenyl)-1H-pyrazol-5-amin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Amino-3-(3-fluorophenyl)pyrazole

AS23824G1 AS23824G5
ACTIAS23824G1EA 0 CHF
ACTIAS23824G1 ACTIAS23824G5
3-(3-Fluorphenyl)-1H-pyrazol-5-amin
3-(3-Fluorphenyl)-1H-pyrazol-5-amin
Formel: C₉H₈FN₃
MW: 177.178 g/mol
MDL Number: MFCD06738760
CAS-Nummer: 766519-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR