1-Boc-piperazin-2-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

AS12156G1 AS12156G5
ACTIAS12156G1EA 0 CHF
ACTIAS12156G1 ACTIAS12156G5
1-Boc-piperazin-2-carbonsäure
1-Boc-piperazin-2-carbonsäure
Formel: C₁₀H₁₈N₂O₄
MDL Number: MFCD02682403
CAS-Nummer: 1214196-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR