2,6-Bis(trifluormethyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Bis(trifluoromethyl)benzeneboronic acid

AS35542G1 AS35542G5
ACTIAS35542G1EA 0 CHF
ACTIAS35542G1 ACTIAS35542G5
2,6-Bis(trifluormethyl)phenylboronsäure
2,6-Bis(trifluormethyl)phenylboronsäure
Formel: C₈H₅BF₆O₂
MW: 257,93 g/mol
MDL Number: MFCD04039321
CAS-Nummer: 681812-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR