Bandage scissors, Lister type
Lieferant: HAMMACHER KARL
HAMMHSB585-18EA 89.1 CHF
HAMMHSB585-18 HAMMHSB583-14 233-1153
Bandage scissors, Lister type
Scheren
These stainless steel scissors are ideal for sizing dressings and removing bandages.
The jaws of the Lister scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without cutting the skin.