Iodmonochlorid solution_SC 0.1 mol/l acc. Wijs, Titripur® Iodine_value, Supelco®
Lieferant: Merck
Gefahr
Synonyms: Iodochloride
MQ100
1.09163.1000 1.09163.2500
1.09163.1000EA 74.05 CHF
1.09163.1000 1.09163.2500
Iodmonochlorid solution_SC 0.1 mol/l acc. Wijs, Titripur® Iodine_value, Supelco®
Iodmonochlorid
Formel: ClI
MW: 162.36 g/mol
Siedepunkt: 97.3
Melting Pt: 26
Dichte: 1.06 g/cm³ (20 °C)
Flash-Pt: 40 °C
MDL Number: MFCD00011354
CAS-Nummer: 7790-99-0
UN: 2789
ADR: 8 (3) II
Merck Index: 13,05039
Order Now
Spezifikation Testergebnisse
Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.