Electroporation cuvettes
Lieferant: BIOZYM
748010 748011 748020 748021 748040 748041
BZYM748010EA 0 CHF
BZYM748010 BZYM748011 BZYM748020 BZYM748021 BZYM748040 BZYM748041
Electroporation cuvettes
Küvetten Elektroporationsküvetten
These gamma sterilised electroporation cuvettes are suitable for all current electroporators.
  • Fitted with tight but easy to open closures
  • Gap width deviation inferior than 0,026 mm
  • Particularly smooth structured cuvette and electrode surfaces