RSLC columns, Dionex™, SolEx™ HRP
Lieferant: DIONEX
074400 075590
DION074400EA 0 CHF
DION074400 DION075590
RSLC columns, Dionex™, SolEx™ HRP
Chromatographiesäulen
Dionex™ SolEx™ HRP columns provide fast, easy sample preconcentration of contaminants in water by solid-phase extraction (SPE) prior to HPLC analysis.
  • Higher retention properties in both polar and hydrophobic analytes
  • No de-wetting, even with 100% aqueous phase
  • High capacity with good recoveries
  • Wide pH compatibility (0 to 14)
  • Wide range of compounds—from polar to hydrophobic analytes
Packed with a hydrophilic reversed phased (HRP) resin comprising divinylbenzene polymer with a hydrophilic bonded layer these columns are placed in the valve to automatically concentrate the analytes of interest.