Cartridges, Dionex™, InGuard™
Lieferant: Thermo Fisher Scientific
Dionex™ InGuard™
DION074034EA 0 CHF
DION074034 DION074035 DION074036 DION074037 DION074038
Cartridges, Dionex™, InGuard™
Chromatographiesäulen
Dionex™ InGuard™ HRP cartridges are ideal for the removal of organic matrix material over a wide range of hydrophobicity including fats from whole milk.
  • Time-saving, in-line sample pretreatment
  • Simplified with automated workflow
  • Reproducible PPM-level determinations in concentrated matrices
  • Increased analytical column lifetimes
  • Leak-free, no-channelling cartridge design
The resin is water-wettable therefore, 100% aqueous samples can be pretreated without disruption of the column bed. Easy installation with standard 10-32 connections. Used singly or in a series with other Dionex InGuard cartridges to suit the application