Hydrometer for Diesel exhaust fluid (DEF)
Lieferant: Bellingham + Stanley
BELL44-862EA 127 CHF
BELL44-862
Hydrometer for Diesel exhaust fluid (DEF)
Aräometer
This hydrometer is specially used to determine whether the concentration of diesel exhaust fluid (DEF) is within limits for use as an additive for selective catalytic reduction SCR of NOx in diesel exhaust fumes.