Overalls with bib and braces for men, C.I., design c, black
Lieferant: Fristads Kansas Group
KWIN870940539C52EA 0 CHF
KWIN870940539C52 KWIN870940539C50 KWIN870940539C56 KWIN870940730C146 KWIN870940440C154 KWIN870940539C54 KWIN870940440C156 KWIN870940440C150 KWIN870940440C152 KWIN870940331C146 KWIN870940331C148 KWIN870940210C48 KWIN870940640C46 KWIN870940210C46 KWIN870940640C48 KWIN870940730C148 KWIN870940539C48 KWIN870940210C44 KWIN870940640C44 KWIN870940440C148 KWIN870940440C48 KWIN870940331C46 KWIN870940440C46 KWIN870940941C50 KWIN870940539C46 KWIN870940440C146 KWIN870940331C48 KWIN870940539C44 KWIN870940440C44 KWIN870940941C52 KWIN870940331C44 KWIN870940231C156 KWIN870940231C154 KWIN870940530C62 KWIN870940530C60 KWIN870940941C44 KWIN870940231C152 KWIN870940941C46 KWIN870940231C150 KWIN870940941C48 KWIN870940520C58 KWIN870940940C154 KWIN870940940C156 KWIN870940440C58 KWIN870940941C60 KWIN870940940C150 KWIN870940941C62 KWIN870940440C56 KWIN870940940C152 KWIN870940520C50 KWIN870940440C54 KWIN870940440C52 KWIN870940520C52 KWIN870940520C54 KWIN870940440C50 KWIN870940520C56 KWIN870940231C44 KWIN870940231C146 KWIN870940231C46 KWIN870940231C48 KWIN870940231C148 KWIN870940520C60 KWIN870940941C54 KWIN870940941C56 KWIN870940941C58 KWIN870940520C48 KWIN870940539C62 KWIN870940539C148 KWIN870940440C62 KWIN870940539C146 KWIN870940210C148 KWIN870940520C44 KWIN870940440C60 KWIN870940539C60 KWIN870940210C146 KWIN870940520C46 KWIN870940530C148 KWIN870940231C56 KWIN870940530C146 KWIN870940231C58 KWIN870940131C146 KWIN870940539C58 KWIN870940231C50 KWIN870940940C146 KWIN870940231C52 KWIN870940940C148 KWIN870940131C148 KWIN870940231C54 KWIN870940131C60 KWIN870940520C152 KWIN870940730C52 KWIN870940520C154 KWIN870940730C54 KWIN870940520C156 KWIN870940210C152 KWIN870940730C56 KWIN870940131C62 KWIN870940730C58 KWIN870940210C150 KWIN870940539C156 KWIN870940210C156 KWIN870940210C154 KWIN870940539C152 KWIN870940520C150 KWIN870940539C154 KWIN870940131C150 KWIN870940530C156 KWIN870940539C150 KWIN870940530C154 KWIN870940131C154 KWIN870940530C152 KWIN870940131C152 KWIN870940530C150 KWIN870940231C60 KWIN870940131C156 KWIN870940231C62 KWIN870940730C60 KWIN870940730C44 KWIN870940131C52 KWIN870940730C46 KWIN870940520C146 KWIN870940131C50 KWIN870940730C48 KWIN870940131C56 KWIN870940520C62 KWIN870940131C54 KWIN870940131C58 KWIN870940520C148 KWIN870940730C50 KWIN870940331C56 KWIN870940940C50 KWIN870940331C58 KWIN870940131C46 KWIN870940331C52 KWIN870940131C44 KWIN870940941C156 KWIN870940331C54 KWIN870940131C48 KWIN870940331C50 KWIN870940941C150 KWIN870940530C56 KWIN870940640C152 KWIN870940640C154 KWIN870940530C54 KWIN870940941C154 KWIN870940530C52 KWIN870940640C156 KWIN870940941C152 KWIN870940530C50 KWIN870940210C62 KWIN870940640C60 KWIN870940210C60 KWIN870940940C44 KWIN870940640C62 KWIN870940940C46 KWIN870940640C150 KWIN870940530C58 KWIN870940940C48 KWIN870940730C62 KWIN870940730C152 KWIN870940730C150 KWIN870940331C150 KWIN870940730C156 KWIN870940940C60 KWIN870940940C62 KWIN870940730C154 KWIN870940331C152 KWIN870940331C154 KWIN870940941C148 KWIN870940941C146 KWIN870940331C156 KWIN870940331C60 KWIN870940331C62 KWIN870940530C44 KWIN870940640C58 KWIN870940210C58 KWIN870940210C56 KWIN870940640C146 KWIN870940640C148 KWIN870940940C52 KWIN870940640C50 KWIN870940210C50 KWIN870940940C54 KWIN870940640C52 KWIN870940640C54 KWIN870940530C48 KWIN870940940C56 KWIN870940210C54 KWIN870940940C58 KWIN870940530C46 KWIN870940640C56 KWIN870940210C52
Overalls with bib and braces for men, C.I., design c, black
Arbeitskleidung
These overalls with bib and braces are made from 65% polyester, 35% cotton, with a soft nap on reverse side.
  • Fly with zip
  • Two chest pockets, one with flap, the other with pen pocket
  • Two front pockets, two back pockets with flap
  • Leg pocket with flap and three small pockets
  • Folding-rule pocket, leg pocket with mobile phone pocket with flap and two small pockets
  • Adjustable waist, side openings
  • Knee pockets that open from the inside and can accommodate height-adjustable kneepads
Weight: 260 g/m²
Basic colour: Black
Bestellinformation: Available with 12 different accent colours.