2-Methyl-5-pyridinboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methylpyridine-5-boronic acid pinacol ester, 6-Methylpyridine-3-boronic acid pinacol ester, 2-(6-Methyl-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methyl-3-pyridylboronic acid Pinacol ester

AS50541G1 AS50541G25 AS50541G5
ACTIAS50541G1EA 29 CHF
ACTIAS50541G1 ACTIAS50541G25 ACTIAS50541G5
2-Methyl-5-pyridinboronsäurepinakolester
2-Methyl-5-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
MW: 219,0878 g/mol
MDL Number: MFCD08669608
CAS-Nummer: 610768-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR