5-(Aminomethyl)-2-brompyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (6-Bromopyridin-3-yl)methanamine

AS31580G1 AS31580G5
ACTIAS31580G1EA 0 CHF
ACTIAS31580G1 ACTIAS31580G5
5-(Aminomethyl)-2-brompyridin
5-(Aminomethyl)-2-brompyridin
Formel: C₆H₇BrN₂
MW: 187,04 g/mol
MDL Number: MFCD06738744
CAS-Nummer: 120740-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR