2-Chlor-6-(tributylstannyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51351G1 AS51351G5
ACTIAS51351G1EA 279 CHF
ACTIAS51351G1 ACTIAS51351G5
2-Chlor-6-(tributylstannyl)pyridin
2-Chlor-6-(tributylstannyl)pyridin
Formel: C₁₇H₃₀ClNSn
MDL Number: MFCD10699155
CAS-Nummer: 263698-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR