5-Fluor-3-pyridinboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Fluoropyridine-5-boronic acid pinacol ester, 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

AS51049G1 AS51049G5
ACTIAS51049G1EA 0 CHF
ACTIAS51049G1 ACTIAS51049G5
5-Fluor-3-pyridinboronsäurepinakolester
5-Fluor-3-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
MDL Number: MFCD07780754
CAS-Nummer: 719268-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR